Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
500 ml, 19/26, 0,25 ± ml
Catalog # 612-0448
Supplier:   SciLabware
Volumetric flasks, class A, blue markings
Catalog # 612-0448
Supplier:  SciLabware
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
500 ml, 19/26, 0,25 ± ml
Price
364,00 kr
Log in for availability
1 * 2 Items
Product Details & Documents
MBL®, borosilicate glass, with PE stopper.
  • Marks and inscriptions in high contrast blue enamel, conformity 'H' mark
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • Calibrated to contain (TC, In)
 : DIN 12664, BS EN ISO 1042
Specifications
  • NS:
    19/26
  • Kapacitet:
    500 ml
  • Seq:
    9
  • Tolerance:
    0,25 ± ml