Accessories
Gelatin filters, wavelength 660 nm, bandpass ± 5 nm
Katalog # 633-0118
Leverandør: SHERWOOD SCIENTIFIC INSTRUMENT
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Gelatin filters, wavelength 660 nm, bandpass ± 5 nm
- Seq:04