Inoculating loops and needles, disposable
Platinøse, 1 µl, hvid, pakket enkeltvis
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Flexible polymer.
- Sterilised by irradiation
- Individually wrapped
- Length: 200 mm
Documents
Certificates
Search by Lot NumberSpecifications
- Pakket:pakket enkeltvis
- Kapacitet:1 µl
- Farve:hvid
- Beskrivelse:Platinøse
- Seq:01