Orders LinkContactUsLinkComponent
Inoculating loops and needles, disposable
Inoculating loops and needles, disposable
Platinøse, 1 µl, hvid, pakket enkeltvis
Catalog # 391-2081
Supplier:   Greiner Bio-One
Inoculating loops and needles, disposable
Catalog # 391-2081
Supplier:  Greiner Bio-One
Inoculating loops and needles, disposable
Inoculating loops and needles, disposable
Platinøse, 1 µl, hvid, pakket enkeltvis
Price
1.210,00 kr
Log in for availability
1 * 600 Items
Product Details & Documents
Flexible polymer.
  • Sterilised by irradiation
  • Individually wrapped
  • Length: 200 mm
Documents
Specifications
  • Pakket:
    pakket enkeltvis
  • Kapacitet:
    1 µl
  • Farve:
    hvid
  • Beskrivelse:
    Platinøse
  • Seq:
    01