Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Preparative sorbents, LiChroprep®

LiChroprep® RP-18, 15 - 25 µm, 03, 10 kg
Katalog # 1.13901.9010
Preparative sorbents, LiChroprep®
Katalog # 1.13901.9010
CAS nummer:  
Preparative sorbents, LiChroprep®

LiChroprep® RP-18, 15 - 25 µm, 03, 10 kg

CAS nummer:  
Price
Spørg for pris
Log ind for tilgængelighed
1 * 10 kg
Product Details & Documents

LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.

  • Reproducible homogeneous silica gel matrix
  • Narrow, well defined particle size distribution for high performance and high permeability
  • Excellent selectivity and efficiency
  • Large number of applications
  • Large batch size
  • Comprehensive regulatory documents are available

LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.

Specifikationer

Specifikationer

  • Partikelstørrelse:
    15 - 25 µm
  • Pakn:
    10 kg
  • Porestørrelse:
    03
  • Beskrivelse:
    LiChroprep® RP-18

Specifikationer