Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Beakers, tall form

600 ml, 150 mm, 80 mm
  213-0036
 :   SciLabware
Beakers, tall form
  213-0036
Beakers, tall form
Beakers, tall form

600 ml, 150 mm, 80 mm

 
1.030,00 kr
   
1 * 10 Items
 

PYREX®, borosilicate glass, with spout.

  • Meget god kemisk resistens
  • Høj temperaturresistens
  • Autoklaverbare

Volumeninddeling og skrivefelt.

Certifications: DIN 12331, ISO 3819

 
  • Udv.Ø:
    80 mm
  • Kapacitet:
    600 ml
  • Højde:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.