Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Rotary evaporating flasks

Rotary evaporating flasks

1000 ml, 29/32 NS, 198 mm
Katalog # 201-8401
Leverandør:   SciLabware
Rotary evaporating flasks
Katalog # 201-8401
Leverandør:  SciLabware
Rotary evaporating flasks
Rotary evaporating flasks

1000 ml, 29/32 NS, 198 mm

Price
548,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

PYREX®, borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Ideal for use with rotary evaporators. Large surface area for rapid evaporation. Robust heavy wall construction for added strength. Florentine shape allows easy removal of solid residues.

Specifikationer
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    1000 ml
  • Samlet højde:
    198 mm