Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for centrifuges

Accessories for centrifuges

Adapter til 1×25 ml rør, 25,2×87 mm
  521-1192
   Lav fremstillingspåvirkning
 :   Hettich
Accessories for centrifuges
  521-1192
 :  Hettich
   Lav fremstillingspåvirkning
Accessories for centrifuges
Accessories for centrifuges

Adapter til 1×25 ml rør, 25,2×87 mm

 
537,00 kr
   
1 * 1 Items
 
 
  • Beskrivelse:
    Adapter til 1×25 ml rør, 25,2×87 mm
  • Seq:
    13
  • Cat. No.:
    521-1192
  • Miljøcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.