Orders LinkContactUsLinkComponent
Passive gas detectors, Dositubes
Hydrogenperoxid (32D), Hydrogen peroxide, 0,5 - 40 ppm
Katalog # 151-0084
Leverandør:   GASTEC
Passive gas detectors, Dositubes
Katalog # 151-0084
Leverandør:  GASTEC
Passive gas detectors, Dositubes
Passive gas detectors, Dositubes
Hydrogenperoxid (32D), Hydrogen peroxide, 0,5 - 40 ppm
Price
486,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
  • Compact, lightweight, and versatile (can be used anywhere)
  • Easy direct measuring and reading
  • No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.
Specifikationer
  • Beskrivelse:
    Hydrogenperoxid (32D)
  • Parameter:
    Hydrogen peroxide
  • Område:
    0,5 - 40 ppm
  • Cat. No.:
    151-0084
  • Måletid:
    1 - 10 h
Accessories
Produktinformation
  • Katalog #
    Parameter
    Område
    Tilgængelighed
    Price
  • Eddikesyre (81D)
    Eddikesyre (81D)
    151-0090
    Acetic acid
    0,5 - 100 ppm
    Log ind for tilgængelighed
    1 * 10 Items - 486,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Holder til Dositube
    Holder til Dositube
    151-0095
    Log ind for tilgængelighed
    1 * 3 Items - 364,00 kr