Clamps for universal platforms
300 ml kolbeklemme, 7 (280×330); 16 (457×457); 20 (457×609) for MaxQ 2000
20 (760×457) and 40 (910×609) clamps for MaxQ 3000
20 (760×457) and 40 (910×609) clamps for MaxQ 3000
Katalog # 444-0309
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Documents
BARN30156BI_TDS_EN
Datablad
Specifikationer
- Til:7 (280×330); 16 (457×457); 20 (457×609) for MaxQ 2000
20 (760×457) and 40 (910×609) clamps for MaxQ 3000 - Beskrivelse:300 ml kolbeklemme
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



