Clamps for universal platforms
125 ml kolbeklemme, 13 (280×330); 32 (457×457); 40 (457×609) for MaxQ 2000
44 (760×457) and 80 (910×609) clamps for MaxQ 3000
44 (760×457) and 80 (910×609) clamps for MaxQ 3000
Katalog # 444-0307
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Specifikationer
- Til:13 (280×330); 32 (457×457); 40 (457×609) for MaxQ 2000
44 (760×457) and 80 (910×609) clamps for MaxQ 3000 - Beskrivelse:125 ml kolbeklemme
- Seq:013



