Clamps for universal platforms
25 ml kolbeklemme, 60 (280×330); 64 (457×457); 80 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000
Katalog # 444-0305
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Specifikationer
- Til:60 (280×330); 64 (457×457); 80 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000 - Beskrivelse:25 ml kolbeklemme
- Seq:011



