Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 100 mm
Katalog # 216-2751
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
- Meget god kemisk resistens
- Høj temperaturresistens
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifikationer
- Ø:100 mm
- Beskrivelse:Watch glass, PYREX® borosilicate glass with firepolished edges
- Seq:07