Orders LinkContactUsLinkComponent
Spatulas
Spatulas
Spatula, lab type, double (C), 200 mm
Katalog # 231-0002
Leverandør:   Agencinox
Spatulas
Katalog # 231-0002
Leverandør:  Agencinox
Spatulas
Spatulas
Spatula, lab type, double (C), 200 mm
Price
195,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
  • Excellent chemical and thermal resistance
  • Autoclavable
Specifikationer
  • Beskrivelse:
    Spatula, lab type, double (C)
  • Længde:
    200 mm