Spatulas
Spatula, semi-micro (D), 140 mm
Katalog # 231-0001
Leverandør: Agencinox
Gå til:
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
- Excellent chemical and thermal resistance
- Autoclavable
Documents
Specifikationer
- Beskrivelse:Spatula, semi-micro (D)
- Længde:140 mm