Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Aluminium seals, lined, WHEATON®

Red, open top, PTFE/red rubber, 11 mm, aluminium
  WHEA224211-06
Aluminium seals, lined, WHEATON®
  WHEA224211-06
Aluminium seals, lined, WHEATON®
Aluminium seals, lined, WHEATON®

Red, open top, PTFE/red rubber, 11 mm, aluminium

 
3.120,00 kr
   
1 * 1.000 Items
 

Open top seal provides for filling or retrieving contents with a syringe, large target diameter.

  • For use with 12×32 mm crimp top vials
  • Choice of liner

They are also ideal for use as closures in chromatography and instrumentation applications.

 
  • Ø:
    11 mm
  • Septum:
    PTFE/red rubber
  • Model:
    Red, open top
  • Materiale:
    aluminium
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.