Dråbeflasker, WHEATON®
15-415, blå
Katalog # 215-4014
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifikationer
- Gevind:15-415
- Seq:27
- Lågfarve:blå