Dråbeflasker, WHEATON®
8-425, hvid
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifikationer
- Gevind:8-425
- Seq:09
- Lågfarve:hvid