Orders LinkContactUsLinkComponent
Formalingskugler
Grinding ball, Glas, 0,75 - 1,00 mm, MM 200/MM 400
Catalog # 412-2917
Supplier:   Retsch GmbH
Formalingskugler
Catalog # 412-2917
Supplier:  Retsch GmbH
Formalingskugler
Formalingskugler
Grinding ball, Glas, 0,75 - 1,00 mm, MM 200/MM 400
Price
560,00 kr
Log in for availability
1 * 0,5 kg
Product Details & Documents
Specifications
  • Ø:
    0,75 - 1,00 mm
  • Beskrivelse:
    Grinding ball
  • Materiale:
    Glas
  • Seq:
    04
  • Til brug med:
    MM 200/MM 400
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-0038
    Formalingskugler
    Zirconiumoxid
    20 mm
    Log in for availability
    1 * 1 Items - 1.090,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-2866
    Formalingskugler
    Wolframcarbid
    10 mm
    Log in for availability
    1 * 1 Items - 457,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-2865
    Formalingskugler
    Rustfrit stål
    10 mm
    Log in for availability
    1 * 1 Items - 32,20 kr
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-0036
    Formalingskugler
    Rustfrit stål
    20 mm
    Log in for availability
    1 * 1 Items - 135,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-2864
    Formalingskugler
    Hærdet stål
    10 mm
    Log in for availability
    1 * 1 Items - 22,90 kr
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-0079
    Formalingskugler
    Agat
    10 mm
    Log in for availability
    1 * 1 Items - 104,00 kr