Orders LinkContactUsLinkComponent
Vials, Vacule®, WHEATON®
10,00 ml, 22×55 mm, Yes, 22-350
Katalog # WHEA651907
Leverandør:   DWK Life Sciences
Vials, Vacule®, WHEATON®
Katalog # WHEA651907
Leverandør:  DWK Life Sciences
Vials, Vacule®, WHEATON®
Vials, Vacule®, WHEATON®
10,00 ml, 22×55 mm, Yes, 22-350
Price
3.370,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents
These vacule vials made of 33 low extractable clear borosilicate glass feature a heavy wall construction designed for lyophilisation and freeze-drying applications.
  • Vials have controlled ID, for accurate stopper fit
  • Screw threads eliminate use of crimp seal
 : Meets USP Type I and ASTM E 438 Type I.
 : Closures for 4 ml vials have to be ordred separately.
 : Vials, 5 and 10 ml are supplied with unlined black phenolic cap.
Specifikationer
  • Kasket inkluderet:
    Yes
  • Cap størrelse:
    22-350
  • Diameter×H:
    22×55 mm
  • Kapacitet:
    10,00 ml
  • Seq:
    03