Cryogenic ampoules, Cryule®, WHEATON®
Cryogenic ampoule, pre-scored, klar, 1,2 ml, 11,8×58 mm, 5 mm
Catalog # 479-1126
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
All glass ampoules that can be hermetically sealed to preserve sample purity and extend sample shelf life. Ideal for standards packaging, environmental and diagnostic standards, lyophilisation and cryogenic storage.
- Tamper-evident seal
- For biological sample preservation in glass
- Can be used in the vapour phase of liquid nitrogen
- Special design allows for storage at low temperatures as well as rapid thawing
Made from low extractable borosilicate glass, conform to USP Type I and ASTM E438 Type I, Class A requirements.
A gold band on the stem of the ampoule indicates that it is pre-scored to facilitate snapping off the top eliminating the need to file the ampoule.
Documents
VWRBRO_007b Biobanking_EU
Product Literature
Specifications
- Diameter×H:11,8×58 mm
- Kapacitet:1,2 ml
- Farve:klar
- Beskrivelse:Cryogenic ampoule, pre-scored
- Seq:02
- Ø udv. top:5 mm