Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings

Volumetric flasks, class A, blue markings

25 ml, 110 mm, 0,040 ± ml
Katalog # 612-0081
Volumetric flasks, class A, blue markings
Katalog # 612-0081
Leverandør:  
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings

25 ml, 110 mm, 0,040 ± ml

Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
MÅLEKOLBE 25ML A CC INDDELT MED KANT 1 * 1 Items Katalog #612-3280

Produktet 612-3280 ligner 612-0081, men kan have en anden producent og/eller emballage.

Product Details & Documents

MBL®, borosilicate glass, with smooth neck.

  • Inscriptions in blue enamel
  • Conformity certified, with batch number

ISO 1042

Certifications: ISO 1042

Specifikationer
  • Kapacitet:
    25 ml
  • Højde:
    110 mm
  • Seq:
    04
  • Tolerance:
    0,040 ± ml
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.