Test cassettes for urine
Cotinine test, 100 ng/ml
Leverandør: SURESCREEN
Gå til:
Product Details & Documents
All the features of a test strip with the added benefit of a sample well and pipette transfer, ideal for small volume samples.
- Clean
- Rapid
- Reliable
Documents
Specifikationer
- Beskrivelse:Cotinine test
- Følsomhed:100 ng/ml