Distillation columns, Hempel
300 mm, 29/32 NS, 29/32 NS
Katalog # LENZ05117304
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours, with cooling coil and cooling mantle. Standard ground cone and jacket. Water supplied through plastic connectors with 8 mm external Ø and GL threaded joint.
DIN 12593
: DIN 12593
Specifikationer
- Konusstr.:29/32 NS
- Kappestr.:29/32 NS
- Brugbar længde:300 mm



