Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Twist-off caps

Twist-off caps

Twist-off caps, for 212 ml jars, 53 mm, hvid
Katalog # PONT010511
Leverandør:   PONT PACKAGING
Twist-off caps
Katalog # PONT010511
Leverandør:  PONT PACKAGING
Twist-off caps

Twist-off caps, for 212 ml jars, 53 mm, hvid

Price
Spørg for pris
Log ind for tilgængelighed
1 * 3.420 Items
Product Details & Documents
Specifikationer
  • Gevind:
    53 mm
  • Farve:
    hvid
  • Beskrivelse:
    Twist-off caps, for 212 ml jars
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.