Engangsørepropper, Bilsom 303
Ørepropper, bulkpakning, gul, S
Katalog # 111-1292
Leverandør: HONEYWELL SAFETY
Gå til:
Product Details & Documents
Formstøbt skumpropper uden PVC, fås i to størrelser for at sikre korrekt og komfortabel pasform.
- Polyurethane foam, without cord
- Conical shape for easy insertion
- Smooth outer surface for better hygiene
- Transparent dispenser for convenience
SNR: 33 dB
EN 352-2
Certifications: EN 352-2.
Packaging: Available bulk packed, as individual pairs or as 400 pairs within the LS-400 dispenser.
Specifikationer
- Farve:gul
- Beskrivelse:Ørepropper, bulkpakning
- Seq:04
- Størrelse:S
Accessories
Produktinformation
- Katalog #FarveTilgængelighedPrice
- Ear plug dispenser for use with earplugsEar plug dispenser for use with earplugsDALL1006203-AMsortLog ind for tilgængelighed1 * 1 Items - 921,00 kr