Membranfiltre, type 154
154, 0,45 µm, 47 mm
SART15406-47-N
PES membrane filters are hydrophilic, have high flow rates thanks to the symmetric structure, have a low non-specific protein adsorption and are chemically resistant over a pH range of 1 to 14. They are recommended for the filtration of aqueous solutions as well for the protein filtration. Furthermore, the low level of extractables makes them suitable for environmental analysis.
- Ø:47 mm
- Porestørrelse:0,45 µm
- Model:154
- Seq:09
- Tykkelse:150 μm
- Cat. No.:SART15406-47-N
- Miljøcertificering:Renewable Energy Certificate
- Flowhastighed for vand:>32 ml/min/cm² (1 bar)
- Environmentally Preferable:Lav fremstillingspåvirkning