Orders LinkContactUsLinkComponent
qPCR-blandinger, ABsolute™
ABsolute™ qPCR ROX (500 nM) Mix, 800×50 µl reactions, 1 KIT
Katalog # 733-0325
Leverandør:   Thermo Fisher Scientific
qPCR-blandinger, ABsolute™
Katalog # 733-0325
CAS nummer:  
qPCR-blandinger, ABsolute™
ABsolute™ qPCR ROX (500 nM) Mix, 800×50 µl reactions, 1 KIT
CAS nummer:  
Price
18.840,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
ABsolute™ qPCR blandinger indeholder alle de nødvendige komponenter til at udføre kvantitativ PCR med undtagelse af skabelon og primere. Der fås udgaver til brug med prober eller med SYBR® Green I inkluderet, med eller uden ROX eller Fluorescein-farver for passiv reference.
ABsolute™ qPCR Mixes
ABsolute™ qPCR Mixes give defined and consistent end-point readings as well as maximum sensitivity. These mixes are therefore the superior choice for qPCR applications that use hybridisation probes:
Enhanced and reproducible end-point values are ideal for SNP analysis
Maximum sensitivity due to the optimised buffer, which allows detection of single copy templates
Robust enough to generate reproducible results from difficult templates such as plant DNA and GC-rich templates
Mixes contain Thermo-Start® DNA Polymerase, which remains completely inactive until fully activated by a 15 minute step at 95 °C, thus ensuring maximum specificity
ABsolute™ qPCR SYBR® Green Mixes
ABsolute™ qPCR SYBR® Green Mixes contain optimal levels of SYBR® Green I dye, in an optimised reaction buffer, to provide a high signal-to-background ratio, which ensures early detection of low copy targets:
High signal-to-background ratio ensures clear detection of low copy number templates
Flexible mix with SYBR® Green and MgCl₂ levels optimised to work for a broad spectrum of templates, with an additional vial of MgCl₂ also provided to optimise reactions involving difficult templates
Mixes contain Thermo-Start® DNA Polymerase to minimise non-specific product formation
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    ABsolute™ qPCR ROX (500 nM) Mix, 800×50 µl reactions
  • Cat. No.:
    733-0325
  • Ansøgning:
    qPCR probe