Dropping funnels, conical shape, without pressure balance
100 ml, 19/26 NS, 19/26 NS
Katalog # 201-0915
Leverandør: Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Katalog # 201-0915
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Screw thread retaining device
- With inscription label, without graduation
- Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifikationer
- Konusstr.:19/26 NS
- Kappestr.:19/26 NS
- Kapacitet:100 ml