Tørrerør, U-formede
180, 18 mm
Katalog # LENZ05339018
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Drying tubes, U-shaped, side tubes with olives (Ext.Ø: 6 mm).
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Ø indv.:18 mm
- Længde:180