Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Recombinant rLysozyme™

Recombinant rLysozyme™

6.000 KU
Katalog # 71110-5
Recombinant rLysozyme™
Katalog # 71110-5
CAS nummer:  
Recombinant rLysozyme™

6.000 KU

CAS nummer:  
Price
4.450,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

rLysozyme™ solution contains a highly purified and stabilised recombinant lysozyme that can be used for lysis of gram negative bacteria, such as E. coli.

Specifikationer

Specifikationer

  • Pakn:
    6.000 KU
  • Cat. No.:
    71110-5
  • Enzym type:
    Recombinant
  • Enzym navn:
    rLysozyme™

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.