Orders LinkContactUsLinkComponent
Recombinant rLysozyme™
1.200 KU
  71110-4
Recombinant rLysozyme™
  71110-4
 :  
Recombinant rLysozyme™
1.200 KU
 :  
1.230,00 kr
1 * 1 Each
rLysozyme™ solution contains a highly purified and stabilised recombinant lysozyme that can be used for lysis of gram negative bacteria, such as E. coli.
  • Pakn:
    1.200 KU
  • Cat. No.:
    71110-4
  • Enzym type:
    Recombinant
  • Enzym navn:
    rLysozyme™