Recombinant rLysozyme™
1.200 KU
71110-4
rLysozyme™ solution contains a highly purified and stabilised recombinant lysozyme that can be used for lysis of gram negative bacteria, such as E. coli.
- Pakn:1.200 KU
- Cat. No.:71110-4
- Enzym type:Recombinant
- Enzym navn:rLysozyme™