Orders LinkContactUsLinkComponent
Sample vials, clear, with screw thread, KIMBLE®
Sample vials, clear, with screw thread, KIMBLE®
Katalog # SCERC40903000F0C2
Leverandør:   DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Katalog # SCERC40903000F0C2
Leverandør:  DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Sample vials, clear, with screw thread, KIMBLE®
Price
1.230,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Screw threaded sample vials made from soda-lime glass, with white PE caps unattached.