Kolber, pæreformede med standardslib
250 ml, 19/26 NS, 200 mm, 85 mm
201-1042
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Hals med standardslib. Hvidt mærkningsfelt.
- Udv.Ø:85 mm
- Kappestr.:19/26 NS
- Kapacitet:250 ml
- Højde:200 mm