Orders LinkContactUsLinkComponent
VWR®, Screw caps
VWR®, Screw caps
65 mm, 1000/1500/2000 ml
  215-5649
VWR®, Screw caps
  215-5649
VWR®, Screw caps
VWR®, Screw caps
65 mm, 1000/1500/2000 ml
129,00 kr
1 * 10 Items
  • Gevind:
    65 mm
  • Til flasker:
    1000/1500/2000 ml
Complementary Products
  •  
    Højde
    Kapacitet
    Hals indv. Ø
    Gevind
    Udv.Ø
  • 215-6007
    247,0 mm
    2000 ml
    50 mm
    65 mm
    120,0 mm
    1 * 1 Items - 60,00 kr
  • 215-6002
    94 mm
    100 ml
    24 mm
    32 mm
    47 mm
    1 * 1 Items - 9,90 kr
  • 215-6005
    206 mm
    1000 ml
    50 mm
    65 mm
    96 mm
    1 * 1 Items - 36,70 kr
  • 215-6001
    77,0 mm
    50 ml
    24 mm
    32 mm
    39,0 mm
    1 * 1 Items - 8,80 kr
  • 215-6004
    152 mm
    500 ml
    39 mm
    50 mm
    78 mm
    1 * 1 Items - 23,90 kr
  • 215-6003
    119 mm
    250 ml
    31 mm
    40 mm
    63 mm
    1 * 1 Items - 12,50 kr