VWR®, Screw caps
50 mm, 500 ml
Katalog # 215-5648
Leverandør: VWR Collection
Product Details & Documents
Specifikationer
- Gevind:50 mm
- Til flasker:500 ml
Complementary Products
Produktinformation
- Katalog #HøjdeKapacitetHals indv. ØGevindUdv.ØTilgængelighedPrice
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



