Sample spoons
Sample spoon, 180 mm
Katalog # BURK5320-0180
Leverandør: BURKLE
Gå til:
Product Details & Documents
Made of AISI 304 (1.4301) stainless steel. This specially shaped spoon reaches almost every corner in containers and prevents samples from trickling out.
- For withdrawing small sample quantities
- Largely chemical resistant (not against acids and alkalis)
Specifikationer
- Beskrivelse:Sample spoon
- Længde:180 mm
- Seq:2