Tripod stands
1,2 kg, 105 mm
Katalog # 241-1109
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Cast iron, varnished.
- With M10 threaded hole
- No support rod
- Slip-proof, rubber feet ensure firm footing
Specifikationer
- Armlængde:105 mm
- Vægt:1,2 kg
- Cat. No.:241-1109