Orders LinkContactUsLinkComponent
Pean clamps
Pean clamps
160 mm
Katalog # 229-0005
Leverandør:   BOCHEM
Pean clamps
Katalog # 229-0005
Leverandør:  BOCHEM
Pean clamps
Pean clamps
160 mm
Price
117,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel. For stopping flow in tube connections.
  • Very good thermal resistance
  • Easy cleaning
  • Safe, simple operation
Documents
Specifikationer
  • Længde:
    160 mm