Pean clamps
160 mm
Katalog # 229-0005
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Stainless steel. For stopping flow in tube connections.
- Very good thermal resistance
- Easy cleaning
- Safe, simple operation
Documents
BOCH2292_TDS_EN
Datablad
Specifikationer
- Længde:160 mm