Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Carbon standards

Carbon standards

Mixed carbon standard, 1000 ppm, 500 mL
Katalog # 701-0049
Leverandør:   REAGECON
Carbon standards
Katalog # 701-0049
Leverandør:  REAGECON
CAS nummer:  
Carbon standards

Mixed carbon standard, 1000 ppm, 500 mL

CAS nummer:  
Price
2.050,00 kr
Log ind for tilgængelighed
1 * 500 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.

These standards are independent, traceable and certified.

Certifications: Accredited to IEC/ISO 17025 and ISO 17034.

Documents

Specifikationer

Specifikationer

  • Koncentration:
    1000 ppm
  • Pakn:
    500 mL
  • Beskrivelse:
    Mixed carbon standard

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.