Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Recombinant rLysozyme™

Recombinant rLysozyme™

300 KU
Katalog # 71110-3
Recombinant rLysozyme™
Katalog # 71110-3
CAS nummer:  
Recombinant rLysozyme™

300 KU

CAS nummer:  
Price
517,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

rLysozyme™ solution contains a highly purified and stabilised recombinant lysozyme that can be used for lysis of gram negative bacteria, such as E. coli.

Specifikationer

Specifikationer

  • Pakn:
    300 KU
  • Cat. No.:
    71110-3
  • Enzym type:
    Recombinant
  • Enzym navn:
    rLysozyme™

Specifikationer