Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chemists spatulas

Chemists spatulas

Flexible, elastic chemists spatula, 190 mm, 105 mm
Katalog # HAMMHSN201-10
Leverandør:   HAMMACHER KARL
Chemists spatulas
Katalog # HAMMHSN201-10
Leverandør:  HAMMACHER KARL
Chemists spatulas
Chemists spatulas

Flexible, elastic chemists spatula, 190 mm, 105 mm

Price
677,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Flexible, elastic chemists spatulas with stainless steel blade.

Specifikationer
  • Beskrivelse:
    Flexible, elastic chemists spatula
  • Længde:
    190 mm
  • Seq:
    01
  • Klinge længde:
    105 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.