n-Hexan, LiChrosolv® for liquid chromatography, Supelco®
Katalog # 1.04391.9185
Leverandør: Merck
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in 'gradient grade' as well as 'isocratic grade'. This minimises the gradient effect of the solvent involved.
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (1013 hPa)
- Smeltende Pt:–94,3 °C
- Tæthed:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- FN:1208
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:203-777-6
Documents
Certifikater
Søg efter partinummerMERC1.04391.2500_TDS_EN ProductPage_104391
Product specification
Merck Pharma Segment Brochure
Product Literature
msds_104391
SDS
Specifikationer
Specifikationer
- Pakn:185 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



