Powder spatulas, micro
140 mm, 5 mm
Katalog # HAMMHWN007-05
Leverandør: HAMMACHER KARL
Gå til:
Product Details & Documents
Single-ended powder spatulas made from WIRONIT steel.
Specifikationer
- Længde:140 mm
- Bladbredde:5 mm
Single-ended powder spatulas made from WIRONIT steel.