Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Katalog # SARS70.3030.100
Marketsource
Leverandør:   SARSTEDT
Pipette tips
Katalog # SARS70.3030.100
Leverandør:  SARSTEDT
Marketsource
Pipette tips
Price
517,58 kr
Log ind for tilgængelighed
1 * 1.920 Items
Product Details & Documents
These high quality tips are suitable for the most common pipettes.
  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.