Orders LinkContactUsLinkComponent
Hand towels, SCOTT®
Hand towels, SCOTT®
SCOTT® Natura, C-fold, 2 ply, hvid, 330×250 mm, 20×140 towels
Katalog # KIMW6810
Leverandør:   KIMBERLY CLARK
Hand towels, SCOTT®
Katalog # KIMW6810
Leverandør:  KIMBERLY CLARK
Hand towels, SCOTT®
Hand towels, SCOTT®
SCOTT® Natura, C-fold, 2 ply, hvid, 330×250 mm, 20×140 towels
Price
1.160,00 kr
Log ind for tilgængelighed
1 * 2.800 Items
Product Details & Documents
Disposable hand towels made from AIRFLEX* (a voluminous, textile-like fabric).
  • High absorption performance, and tear-proof even when wet
  • Single, self-presenting towels for enhanced hygiene and less waste
  • Extremely soft
Specifikationer
  • Pakket:
    20×140 towels
  • Farve:
    hvid
  • Beskrivelse:
    SCOTT® Natura, C-fold, 2 ply
  • L×B:
    330×250 mm
  • Seq:
    03
Accessories
Produktinformation
  • Katalog #
    B×D×H
    Tilgængelighed
    Price
  • Dispenser for hand towel rolls, AQUARIUS* 6959, white ABS, with window
    Dispenser for hand towel rolls, AQUARIUS* 6959, white ABS, with window
    115-1779
    326×241×430 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.480,00 kr