Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, stainless steel, 600 mm, I.Ø 0,5 mm, to connect autosampler to column
  HEWLG2260-87300
Capillaries for LC systems
  HEWLG2260-87300
Capillaries for LC systems
Capillaries for LC systems
Capillaries, stainless steel, 600 mm, I.Ø 0,5 mm, to connect autosampler to column
806,00 kr
1 * 1 Items
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
  • Beskrivelse:
    Capillaries, stainless steel, 600 mm, I.Ø 0,5 mm, to connect autosampler to column
  • Seq:
    62