Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries, stainless steel, 160 mm, I.Ø 0,5 mm, to connect injector valve to prep head
Katalog # HEWLG2258-87301
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG2258-87301
Capillaries for LC systems
Capillaries for LC systems
Capillaries, stainless steel, 160 mm, I.Ø 0,5 mm, to connect injector valve to prep head
Price
776,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, stainless steel, 160 mm, I.Ø 0,5 mm, to connect injector valve to prep head
  • Seq:
    61