Capillaries for LC systems
Capillaries, stainless steel, 160 mm, I.Ø 0,5 mm, to connect injector valve to prep head
Katalog # HEWLG2258-87301
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, stainless steel, 160 mm, I.Ø 0,5 mm, to connect injector valve to prep head
- Seq:61