Capillaries for LC systems
Capillaries, blue, stainless steel, 120 mm, I.Ø 0,25 mm, to connect injector valve to waste
Katalog # HEWLG1377-87301
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, blue, stainless steel, 120 mm, I.Ø 0,25 mm, to connect injector valve to waste
- Seq:19