Capillaries for LC systems
Capillaries, A/A, stainless steel, to connect filter to EMPV
Katalog # HEWLG1375-87400
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, A/A, stainless steel, to connect filter to EMPV
- Seq:18
- Cat. No.:HEWLG1375-87400