Capillaries for LC systems
Capillaries, PEEK coated fused silica, C/D, 500 mm, I.Ø 1905 mm, to connect injection valve to column
Catalog # HEWLG1375-87311
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
- Beskrivelse:Capillaries, PEEK coated fused silica, C/D, 500 mm, I.Ø 1905 mm, to connect injection valve to column
- Seq:112